Benzoic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.22257.0100
8.22257.1000
8.22257.5000
8.22257.0100
8.22257.1000
8.22257.5000
EM8.22257.0100CS
3573.96
CAD
EM8.22257.0100
EM8.22257.1000
EM8.22257.5000
CA8.22257.0100
CA8.22257.1000
CA8.22257.5000
Benzoic acid for synthesis, Sigma-Aldrich®
Benzoic acid
Formula:
C₇H₆O₂ Boiling Pt: 249 °C (1013 hPa) Melting Pt: 121…123 °C Density: 1.316 g/cm³ (20 °C) Flash Pt: 121 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00002398 CAS Number: 65-85-0 EINECS: 200-618-2 REACH: 01-2119455536-33 Merck Index: 13,01092 |
|
Specification Test Results
Assay (acidimetric) | ≥ 99.0 % |
Identity (IR) | passes test |
Melting range (lower value) | ≥ 122 °C |
Melting range (upper value) | ≤ 125 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...