Tween® 40 (Polysorbate) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Polyoxyethylene sorbitan monopalmitate

M-Clarity™ quality level = MQ200

8.22185.0500 8.22185.1000 8.22185.0500 8.22185.1000
EM8.22185.0500CS 3129.98 CAD
EM8.22185.0500 EM8.22185.1000 CA8.22185.0500 CA8.22185.1000
Tween® 40 (Polysorbate) for synthesis, Sigma-Aldrich®
Tween® 40 (Polysorbate)
Formula: C₆₂H₁₂₂O₂₆
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: ∼0.1 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00165345
CAS Number: 9005-66-7

Order Now

Specification Test Results

Solubility (20 °C) soluble
Boiling Point 100 °C
Flash Point 149 °C
Density 1.09 g/cm3 (20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR