Tetramethylammonium chloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.22156.0250
8.22156.1000
8.22156.5000
8.22156.0250
8.22156.1000
8.22156.5000
EM8.22156.0250CS
701.52
CAD
EM8.22156.0250
EM8.22156.1000
EM8.22156.5000
CA8.22156.0250
CA8.22156.1000
CA8.22156.5000
Tetramethylammonium chloride for synthesis, Sigma-Aldrich®
Tetramethylammonium chloride
Formula:
(CH₃)₄NCl Melting Pt: > 300 °C Density: 1.169 g/cm³ (20 °C) Flash Pt: >100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011628 CAS Number: 75-57-0 EINECS: 200-880-8 UN: 2811 ADR: 6.1,II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point |
|
Molar Mass | 109.60 g/mol |
Vapor Pressure | (20 °C) |
Flash Point |
|
Density | 1.17 g/cm3 (20 °C) |
pH | 6 - 8 (100 g/l, H2O, 20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...