Tetraethylammonium bromide for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
M-Clarity™ quality level = MQ200
8.22147.0250
8.22147.1000
8.22147.0250
8.22147.1000
EM8.22147.0250CS
706.26
CAD
EM8.22147.0250
EM8.22147.1000
CA8.22147.0250
CA8.22147.1000
Tetraethylammonium bromide for synthesis, Sigma-Aldrich®
Tetraethylammonium bromide
Formula:
C₈H₂₀BrN Melting Pt: > 285 °C Density: 1.43 g/cm³ (20 °C) |
MDL Number:
MFCD00011825 CAS Number: 71-91-0 EINECS: 200-769-4 Merck Index: 13,09274 |
Specification Test Results
Solubility | 2795 g/l (25 °C) |
Melting Point |
|
Molar Mass | 210.16 g/mol |
Density | 1.397 g/cm3 (20 °C) |
pH | 6.5 (100 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...