Propionaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Propanal

M-Clarity™ quality level = MQ200

8.22133.0100 8.22133.1000 8.22133.0100 8.22133.1000
EM8.22133.0100CS 1464.66 CAD
EM8.22133.0100 EM8.22133.1000 CA8.22133.0100 CA8.22133.1000
Propionaldehyde for synthesis, Sigma-Aldrich®
Propionaldehyde
Formula: C₃H₆O
MW: 58.08 g/mol
Boiling Pt: 48 °C (1013 hPa)
Melting Pt: –81 °C
Density: 0.811 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007020
CAS Number: 123-38-6
EINECS: 204-623-0
UN: 1275
ADR: 3,II
Merck Index: 13,07915

Order Now

Specification Test Results

Solubility 200 g/l (20 °C)
Melting Point -81 °C
Molar Mass 58.08 g/mol
Boiling Point 47 - 48 °C (1013 hPa)
Vapor Pressure 343 hPa (20 °C)
Flash Point -40 °C
Refractive Index 1.365 (20 °C, 589 nm)
Explosion Limit 2.3 - 21 %(V)
Density 0.797 g/cm3 (20 °C)
Ignition Point 175 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR