Sodium dodecyl sulfate (SDS) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

M-Clarity™ quality level = MQ200

8.22050.1000 8.22050.0100 8.22050.2500 8.22050.0100 8.22050.2500
CA8.22050.1000EA 1139 CAD
CA8.22050.1000 EM8.22050.0100 EM8.22050.2500 CA8.22050.0100 CA8.22050.2500
Sodium dodecyl sulfate (SDS) for synthesis, Sigma-Aldrich®
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: 204…207 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 150 g/l (20 °C)
Melting Point 204.06 - 207.06 °C
Molar Mass 288.4 g/mol
Bulk Density 490 - 560 kg/m3
Vapor Pressure (20 °C)
Flash Point 150 °C
Density 1.1 g/cm3 (20 °C)
pH 6 - 9 (10 g/l, H2O, 20 °C)
Hazard Class 4.1
Hazard Class Description Flammable Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR