4-(Dimethylamino)cinnamaldehyde for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: p-Dimethylamino-Cinnamaldehyde

M-Clarity™ quality level = MQ200

8.22034.0002 8.22034.0010 8.22034.0500 8.22034.0002 8.22034.0010 8.22034.0500
EM8.22034.0002CS 11538.74 CAD
EM8.22034.0002 EM8.22034.0010 EM8.22034.0500 CA8.22034.0002 CA8.22034.0010 CA8.22034.0500
4-(Dimethylamino)cinnamaldehyde for synthesis, Sigma-Aldrich®
4-(Dimethylamino)cinnamaldehyde
Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD00007002
CAS Number: 6203-18-5
EINECS: 228-267-0

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 135 - 138 °C
Molar Mass 175.22 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR