Polyethylene glycol 10,000 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.21881.1000 8.21881.1000 8.21881.5000
EM8.21881.1000CS 813.7 CAD
EM8.21881.1000 CA8.21881.1000 CA8.21881.5000
Polyethylene glycol 10,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 10,000
Formula: H(OCH₂CH₂)nOH
Melting Pt: 63 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 265 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 58 - 63 °C
Molar Mass 9000 g/mol
Bulk Density 400 - 500 kg/m3
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 265 °C
Density 1.2 g/cm3 (20 °C)
pH 5 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 340 °C
Viscosity kinematic 500 - 900 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR