4-Hydroxybenzoic acid for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.21814.0005
8.21814.0250
8.21814.1000
8.21814.0005
8.21814.0250
8.21814.1000
EM8.21814.0005CS
34905.36
CAD
EM8.21814.0005
EM8.21814.0250
EM8.21814.1000
CA8.21814.0005
CA8.21814.0250
CA8.21814.1000
4-Hydroxybenzoic acid for synthesis, Sigma-Aldrich®
4-Hydroxybenzoic acid
Formula:
C₇H₆O₃ Boiling Pt: 336 °C (1013 hPa) Melting Pt: 210…211 °C Density: 1.398 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002547 CAS Number: 99-96-7 EINECS: 202-804-9 Merck Index: 13,04837 |
Specification Test Results
Assay (HPLC, area%) | ≥ 98.0 % (a/a) |
Identity (IR) | passes test |
Melting range (lower value) | ≥ 214 °C |
Melting range (upper value) | ≤ 217 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...