(±)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DL-1-Phenylethylamine, (±)-1-Phenylethylamine, (±)-α-Phenylethylamine

M-Clarity™ quality level = MQ200

8.21256.0500 8.21256.0100 8.21256.0500
EM8.21256.0100CS 1167.62 CAD
EM8.21256.0100 EM8.21256.0500 CA8.21256.0100 CA8.21256.0500
(±)-α-Methylbenzylamine for synthesis, Sigma-Aldrich®
(±)-α-Methylbenzylamine
Formula: C₆H₅CH(NH₂)CH₃
MW: 121.18 g/mol
Boiling Pt: 184…186 °C (1013 hPa)
Melting Pt: –65 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00008069
CAS Number: 618-36-0
EINECS: 210-545-8
UN: 2735
ADR: 8 II
Merck Index: 12,06106
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7749b7c7d0770d82ae5145b1a236458393427163&vid=9ef2ef0e518b58972043eaec1de61438a07bcc37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 42 g/l (20 °C)
Melting Point -65 °C
Molar Mass 121.18 g/mol
Boiling Point 184 - 186 °C (1013 hPa)
Vapor Pressure 0.7 hPa (20 °C)
Flash Point 70 °C
Refractive Index 1.5253 (20 °C, 589 nm)
Density 0.95 g/cm3 (20 °C)
pH 7 (H2O, 20 °C)
Ignition Point 355 °C
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR