Hydroquinone monomethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.21233.0005 8.21233.0025 8.21233.0250 8.21233.0005 8.21233.0025 8.21233.0250
EM8.21233.0005CS 34165.92 CAD
EM8.21233.0005 EM8.21233.0025 EM8.21233.0250 CA8.21233.0005 CA8.21233.0025 CA8.21233.0250
Hydroquinone monomethyl ether for synthesis, Sigma-Aldrich®
Hydroquinone monomethyl ether
Formula: C₇H₈O₂
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 55…57 °C
Density: 1.55 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8

Order Now

Specification Test Results

Solubility 40 g/l (25 °C)
Melting Point 54 - 57 °C
Molar Mass 124.14 g/mol
Boiling Point 243 °C (1013 hPa)
Vapor Pressure 0.13 hPa (60 °C)
Flash Point 133 °C
Density 1.25 g/cm3 (20 °C)
pH 5.1 (30 g/l, H2O, 20 °C)
Ignition Point 445 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR