Trimethyl borate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.21180.0250 8.21180.1000 8.21180.0250 8.21180.1000
EM8.21180.0250CS 979.6 CAD
EM8.21180.0250 EM8.21180.1000 CA8.21180.0250 CA8.21180.1000
Trimethyl borate for synthesis, Sigma-Aldrich®
Trimethyl borate
Formula: C₃H₉BO₃
Boiling Pt: 67 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.915 g/cm³ (20 °C)
Flash Pt: –11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008346
CAS Number: 121-43-7
EINECS: 204-468-9
UN: 2416
ADR: 3,II
Merck Index: 12,09842

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point -34 - -29 °C
Molar Mass 103.91 g/mol
Boiling Point 67 - 69 °C (1013 hPa)
Vapor Pressure 182 hPa (25 °C)
Flash Point -8 °C
Refractive Index 1.3568 (20 °C)
Explosion Limit 0.932 g/cm3 (20 °C)
Density (H2O) no data available
Ignition Point 305 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR