N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
N1,N1,N4,N4-Tetramethylbenzene-1,4-diamine dihydrochloride, TMPD 2 HCl, N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride, N,N,N',N'-Tetramethyl-1,4-phenylenediammonium dichloride
M-Clarity™ quality level = MQ200
8.21102.0010
8.21102.0025
8.21102.0010
8.21102.0025
EM8.21102.0010CS
18481.26
CAD
EM8.21102.0010
EM8.21102.0025
CA8.21102.0010
CA8.21102.0025
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride for synthesis, Sigma-Aldrich®
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
Formula:
C₁₀H₁₈Cl₂N₂ MW: 237.17 g/mol Melting Pt: 227…230 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012482 CAS Number: 637-01-4 EINECS: 211-274-8 Merck Index: 12,09367 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 227 - 230 °C (decomposition) |
Molar Mass | 237.17 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...