Glycofural for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Glycofurol, Tetrahydrofurfuryl alcohol polyethylene glycol ether
M-Clarity™ quality level = MQ200
8.21092.0100
8.21092.0250
8.21092.0100
8.21092.0250
EM8.21092.0100CS
2074.54
CAD
EM8.21092.0100
EM8.21092.0250
CA8.21092.0100
CA8.21092.0250
Glycofural for synthesis, Sigma-Aldrich®
Glycofural
Formula:
C₇H₁₄O₃ MW: 146.186 g/mol |
MDL Number:
MFCD00801146 CAS Number: 31692-85-0 |
Specification Test Results
Solubility | (20 °C) soluble |
Boiling Point | 176 °C (40 hPa) |
Refractive Index | 1.458 (27 °C, 589 nm) |
Density | 1.08 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...