Polyvinyl alcohol, M.W. ∼ 30000 for synthesis, fully hydrolysed, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Mowiol®Poly(vinyl alcohol)PVOHPVA

M-Clarity™ quality level = MQ200

8.21039.1000 8.21039.0100 8.21039.1000 8.21039.0100
CA8.21039.0100CS 1087.7 CAD
CA8.21039.0100 CA8.21039.1000 EM8.21039.0100 EM8.21039.1000
Polyvinyl alcohol, M.W. ∼ 30000 for synthesis, fully hydrolysed, Sigma-Aldrich®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Density 1.3 g/cm3 (20 °C)
pH 4.5 - 7 (40 g/l, H2O, 20 °C)
Ignition Point 450 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR