Phosphorus pentasulfide for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.21024.0100 8.21024.1000 8.21024.2500 8.21024.0100 8.21024.1000 8.21024.2500
EM8.21024.0100CS 13937.18 CAD
EM8.21024.0100 EM8.21024.1000 EM8.21024.2500 CA8.21024.0100 CA8.21024.1000 CA8.21024.2500
Phosphorus pentasulfide for synthesis, Sigma-Aldrich®
Phosphorus pentasulphide
Formula: P₂S₅
MDL Number: MFCD00011441
CAS Number: 1314-80-3
UN: 1340
ADR: 4.3 (4.1),II

Order Now

Specification Test Results

Solubility (20 °C) (decomposition)
Melting Point 284 - 287 °C
Molar Mass 222.28 g/mol
Bulk Density 900 - 1100 kg/m3
Boiling Point 513 - 515 °C (1013 hPa)
Density 2.08 g/cm3 (20 °C)
pH 1 (10 g/l, H2O, 20 °C)
Ignition Point 142 °C
Hazard Class 4.3
Hazard Class Description Dangerous When Wet Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR