2-(2-Butoxyethoxy)ethyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Diethyleneglycolmonobutyletheracetate, Diethylene glycol monobutyl ether acetate

M-Clarity™ quality level = MQ200

8.21014.0100 8.21014.1000 8.21014.2500 8.21014.0100 8.21014.1000 8.21014.2500
EM8.21014.0100CS 701.52 CAD
EM8.21014.0100 EM8.21014.1000 EM8.21014.2500 CA8.21014.0100 CA8.21014.1000 CA8.21014.2500
2-(2-Butoxyethoxy)ethyl acetate for synthesis, Sigma-Aldrich®
2-(2-Butoxyethoxy)ethyl acetate
Formula: C₁₀H₂₀O₄
MW: 204.27 g/mol
Boiling Pt: 246
Melting Pt: -32
Flash Pt: 102
MDL Number: MFCD00009458
CAS Number: 124-17-4

Order Now

Specification Test Results

Solubility 64 g/l (20 °C)
Melting Point -32 °C
Molar Mass 204.26 g/mol
Boiling Point 244 - 250 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 102 °C
Refractive Index 1.4262 (20 °C, 589 nm)
Explosion Limit 1.0 - 5.3 %(V)
Density 0.98 g/cm3 (20 °C)
pH 3 - 4 (50 g/l, H2O, 20 °C)
Ignition Point 290 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR