m-Phenylenediamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: MPD, 1,3-Diaminobenzene, 1,3-Phenylenediamine

M-Clarity™ quality level = MQ200

8.20991.0005 8.20991.0250 8.20991.1000 8.20991.0005 8.20991.0250 8.20991.1000
EM8.20991.0005CS 45815.26 CAD
EM8.20991.0005 EM8.20991.0250 EM8.20991.1000 CA8.20991.0005 CA8.20991.0250 CA8.20991.1000
m-Phenylenediamine for synthesis, Sigma-Aldrich®
m-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 282
Flash Pt: 187
MDL Number: MFCD00007799
CAS Number: 108-45-2
UN: 1673
ADR: 6.1 III
Merck Index: 12,07437

Order Now

Specification Test Results

Solubility 350 g/l (25 °C)
Melting Point 63 - 64 °C
Molar Mass 108.14 g/mol
Bulk Density 600 kg/m3
Boiling Point 283 - 284 °C (1013 hPa)
Vapor Pressure 1.3 hPa (100 °C)
Flash Point 187 °C
Density 1.14 g/cm3 (20 °C)
pH 8 (100 g/l, H2O, 20 °C)
Ignition Point 560 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR