Diphenyl ether zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Diphenyl oxide, Phenyl ether, Phenoxybenzene

MQ200

8.20978.0100 8.20978.1000 8.20978.2500 8.20978.0100 8.20978.1000 8.20978.2500
EM8.20978.0100CS 1289.28 CAD
EM8.20978.0100 EM8.20978.1000 EM8.20978.2500 CA8.20978.0100 CA8.20978.1000 CA8.20978.2500
Diphenyl ether zS, Sigma-Aldrich®
Diphenyl ether
Formula: C₁₂H₁₀O
MW: 170.21 g/mol
Boiling Pt: 498
Flash Pt: 125
MDL Number: MFCD00003034
CAS Number: 101-84-8
UN: 3077
ADR: 9 III
Merck Index: 12,07442

Order Now

Specification Test Results

Solubility 0.018 g/l (25 °C)
Melting Point 24 - 27 °C
Molar Mass 170.21 g/mol
Boiling Point 259 °C (1013 hPa)
Vapor Pressure 0.05 hPa (20 °C)
Flash Point 115 °C
Refractive Index 1.5785 (25 °C, 589 nm)
Explosion Limit 0.8 - 1.5 %(V)
Density 1.07 g/cm3 (20 °C)
Ignition Point 610 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR