Tetraethylene glycol dimethyl ether for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20959.0250 8.20959.1000 8.20959.0250 8.20959.1000
EM8.20959.0250CS 6332.64 CAD
EM8.20959.0250 EM8.20959.1000 CA8.20959.0250 CA8.20959.1000
Tetraethylene glycol dimethyl ether for synthesis, Sigma-Aldrich®
Tetraethylene glycol dimethyl ether
Formula: C₁₀H₂₂O₅
MDL Number: MFCD00008505
CAS Number: 143-24-8

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -30 °C
Molar Mass 222.28 g/mol
Boiling Point 272 - 277 °C (1013 hPa)
Vapor Pressure 0.01 hPa (20 °C)
Flash Point 141 °C
Refractive Index 1.4322 (20 °C, 589 nm)
Explosion Limit 0.8 %(V)
Density 1.01 g/cm3 (20 °C)
Ignition Point 200 °C
Viscosity kinematic 4.1 mm2/s (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR