1-Octanol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
n-Octanol
M-Clarity™ quality level = MQ200
8.20931.0100
8.20931.1000
8.20931.2500
8.20931.0100
8.20931.1000
8.20931.2500
EM8.20931.0100CS
1295.6
CAD
EM8.20931.0100
EM8.20931.1000
EM8.20931.2500
CA8.20931.0100
CA8.20931.1000
CA8.20931.2500
1-Octanol for synthesis, Sigma-Aldrich®
1-Octanol
Formula:
H₃C(CH₂)₆CH₂OH MW: 130.23 g/mol |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 |
Specification Test Results
Solubility | 0.3 g/l (20 °C) |
Melting Point | -16 °C |
Molar Mass | 130.23 g/mol |
Boiling Point | 195 °C (1013 hPa) |
Vapor Pressure | 0.031 hPa (20 °C) |
Flash Point | 90 °C |
Explosion Limit | 0.8 %(V) |
Density | 0.83 g/cm3 (20 °C) |
pH | (H2O) not applicable |
Ignition Point | 270 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...