1-Octene for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20928.0005
8.20928.0250
8.20928.1000
8.20928.0005
8.20928.0250
8.20928.1000
EM8.20928.0005CS
4869.56
CAD
EM8.20928.0005
EM8.20928.0250
EM8.20928.1000
CA8.20928.0005
CA8.20928.0250
CA8.20928.1000
1-Octene for synthesis, Sigma-Aldrich®
1-Octene
Formula:
C₈H₁₆ MW: 112.22 g/mol Boiling Pt: 121 Melting Pt: -102 Flash Pt: 8 |
MDL Number:
MFCD00009548 CAS Number: 111-66-0 UN: 3295 ADR: 3 II Merck Index: 13,01770 |
Specification Test Results
Solubility | (20 °C) practically insoluble |
Melting Point | -102 °C |
Molar Mass | 112.21 g/mol |
Boiling Point | 121 °C |
Vapor Pressure | 17.3 hPa (20 °C) |
Flash Point | 8 °C |
Refractive Index | 1.4085 (20 °C, 589 nm) |
Explosion Limit | 0.7 - 6.8 %(V) |
Density | 0.72 g/cm3 (20 °C) |
Ignition Point | 221 °C |
Viscosity kinematic | 0.38 mm2/s (40 °C) |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...