Naphthalene for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.20846.0100 8.20846.1000 8.20846.0100 8.20846.1000
EM8.20846.0100CS 2562.76 CAD
EM8.20846.0100 EM8.20846.1000 CA8.20846.0100 CA8.20846.1000
Naphthalene for synthesis, Sigma-Aldrich®
Naphthalene
Formula: C₁₀H₈
MDL Number: MFCD00001742
CAS Number: 91-20-3
UN: 1334
ADR: 4.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.03 g/l (25 °C)
Melting Point 79 - 82 °C
Molar Mass 128.17 g/mol
Bulk Density 550 - 600 mg/m3
Boiling Point 218 °C
Vapor Pressure 0.08 hPa (20 °C)
Refractive Index 80 °C
Explosion Limit 0.9 - 5.9 %(V)
Density 1.15 g/cm3 (20 °C)
Ignition Point 540 °C
Hazard Class 4.1
Hazard Class Description Flammable Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR