Dimethyl sulfide ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: DMS

M-Clarity™ quality level = MQ200

8.20833.1000 8.20833.0005 8.20833.0250 8.20833.1000 8.20833.9045
EM8.20833.0005CS 6863.52 CAD
EM8.20833.0005 EM8.20833.0250 EM8.20833.1000 CA8.20833.0005 CA8.20833.0250 CA8.20833.1000 CA8.20833.9045
Dimethyl sulfide ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Dimethyl sulphide
Formula: C₂H₆S
MW: 62.14 g/mol
Boiling Pt: 37 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.85 g/cm3 (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00008562
CAS Number: 75-18-3
UN: 1164
ADR: 3,II

Order Now

Specification Test Results

Solubility 2 g/l (20 °C)
Melting Point -98 °C
Molar Mass 62.14 g/mol
Boiling Point 37 °C (1013 hPa)
Vapor Pressure 530 hPa (25 °C)
Flash Point -48 °C
Refractive Index 1.4351 (20 °C, 589 nm)
Explosion Limit 2.2 - 19.7 %(V)
Density 0.85 g/cm3 (20 °C)
Ignition Point 205 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR