2-Methylfuran stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20798.0250 8.20798.1000 8.20798.0250 8.20798.1000
EM8.20798.0250CS 1192.9 CAD
EM8.20798.0250 EM8.20798.1000 CA8.20798.0250 CA8.20798.1000
2-Methylfuran stabilized for synthesis, Sigma-Aldrich®
2-Methylfuran
Formula: C₅H₆O
MW: 82.1 g/mol
MDL Number: MFCD00003248
CAS Number: 534-22-5
UN: 2301
ADR: 3,II

Order Now

Specification Test Results

Solubility 3000 mg/l (20 °C)
Melting Point -89 °C
Molar Mass 82.10 g/mol
Boiling Point 63 - 66 °C (1013 hPa)
Vapor Pressure 139 hPa (20 °C)
Flash Point -27 °C
Refractive Index 1.4332 (20 °C, 589 nm)
Density 0.91 g/cm3 (20 °C)
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR