n-Hexadecane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Hexadecane, Cetane

M-Clarity™ quality level = MQ200

8.20633.0005 8.20633.0250 8.20633.1000 8.20633.0005 8.20633.0250 8.20633.1000
EM8.20633.0005CS 2908.78 CAD
EM8.20633.0005 EM8.20633.0250 EM8.20633.1000 CA8.20633.0005 CA8.20633.0250 CA8.20633.1000
n-Hexadecane for synthesis, Sigma-Aldrich®
n-Hexadecane
Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
MDL Number: MFCD00008998
CAS Number: 544-76-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 18 °C
Molar Mass 226.44 g/mol
Boiling Point 287 °C
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 135 °C
Density 0.77 g/cm3 (20 °C)
Viscosity kinematic  - 7 mm2/s (40 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR