Guanidinium thiocyanate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Guanidinium sulfocyanide, Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide

M-Clarity™ quality level = MQ200

8.20613.0250 8.20613.1000 8.20613.0250 8.20613.1000
EM8.20613.0250CS 3389.1 CAD
EM8.20613.0250 EM8.20613.1000 CA8.20613.0250 CA8.20613.1000
Guanidinium thiocyanate for synthesis, Sigma-Aldrich®
Guanidinium thiocyanate
Formula: CH₅N₃·CHNS
MW: 118.16 g/mol
Melting Pt: 118
MDL Number: MFCD00013027
CAS Number: 593-84-0
UN: 1759
ADR: 8 III

Order Now

Specification Test Results

Solubility 1420 g/l (20 °C)
Melting Point 118 °C
Molar Mass 118.16 g/mol
Bulk Density 630 kg/m3
Density 1.29 g/cm3 (20 °C)
pH 4.8 - 6.0 (1420 g/l, H2O, 20 °C) (saturated solution)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR