Glyoxal 40% in water for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Ethandial

M-Clarity™ quality level = MQ200

8.20610.2500 8.20610.9025
CA8.20610.2500CS 661 CAD
CA8.20610.2500 CA8.20610.9025
Glyoxal 40% in water for synthesis, Sigma-Aldrich®
Glyoxal
Formula: C₂H₂O₂
MW: 58.04 g/mol
MDL Number: MFCD00006957
CAS Number: 107-22-2

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -14 °C
Boiling Point 104 °C (1013 hPa)
Vapor Pressure 20.2 hPa (20 °C)
Flash Point 84 °C
Refractive Index 1.3826 (20 °C, 589 nm)
Density 1.27 g/cm3 (20 °C)
pH 2 - 3.5 (400 g/l, H2O, 20 °C)
Ignition Point 285 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR