Isopropyl acetate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid isopropyl ester

M-Clarity™ quality level = MQ200

8.20560.0100 8.20560.1000 8.20560.2500 8.20560.0100 8.20560.1000 8.20560.2500
EM8.20560.0100CS 1014.36 CAD
EM8.20560.0100 EM8.20560.1000 EM8.20560.2500 CA8.20560.0100 CA8.20560.1000 CA8.20560.2500
Isopropyl acetate for synthesis, Sigma-Aldrich®
Isopropyl acetate
Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD00008877
CAS Number: 108-21-4
UN: 1220
ADR: 3,II

Order Now

Specification Test Results

Solubility 31 g/l (20 °C)
Melting Point -73 °C
Molar Mass 102.13 g/mol
Boiling Point 88 - 90 °C (1013 hPa)
Vapor Pressure 61 hPa (20 °C)
Flash Point 1.8 - 8 %(V)
Refractive Index 4 °C
Density 0.87 g/cm3 (20 °C)
Ignition Point 460 °C
Hazard Class 3
Hazard Class Description Flammable Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR