Diphenylamine for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: N-Phenylaniline, N-Phenylbenzeneamine

M-Clarity™ quality level = MQ200

8.20528.0100 8.20528.1000 8.20528.0100 8.20528.1000
EM8.20528.0100CS 3267.44 CAD
EM8.20528.0100 EM8.20528.1000 CA8.20528.0100 CA8.20528.1000
Diphenylamine for synthesis, Sigma-Aldrich®
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302
Flash Pt: 153
MDL Number: MFCD00003014
CAS Number: 122-39-4
UN: 3077
ADR: 9 III
Merck Index: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.05 g/l (25 °C)
Melting Point 53 - 54 °C
Molar Mass 169.23 g/mol
Bulk Density 610 kg/m3
Boiling Point 302 °C (1013 hPa)
Vapor Pressure 0.0003 hPa (20 °C)
Flash Point 153 °C DIN 51758
Density 1.16 g/cm3 (20 °C)
Ignition Point 630 °C DIN 51794
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR