1,3-Dihydroxyacetone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.20482.0100 8.20482.0250 8.20482.0100 8.20482.0250
EM8.20482.0100CS 2463.22 CAD
EM8.20482.0100 EM8.20482.0250 CA8.20482.0100 CA8.20482.0250
1,3-Dihydroxyacetone for synthesis, Sigma-Aldrich®
1,3-Dihydroxyacetone
Formula: C₃H₆O₃
MDL Number: MFCD00004670
CAS Number: 96-26-4

Order Now

Specification Test Results

Solubility 250 g/l (20 °C)
Melting Point 83 °C Monomer
Molar Mass 90.07 g/mol
Boiling Point 188 °C (decomposition),OECD Test Guideline 103
Vapor Pressure  - 0.01 Pa (20 °C)
Density 1.52 g/cm3 (20 °C) OECD Test Guideline 109
pH 4 - 6 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR