1,2-Diethoxyethane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20390.0050 8.20390.0250 8.20390.0050 8.20390.0250
EM8.20390.0050CS 3581.86 CAD
EM8.20390.0050 EM8.20390.0250 CA8.20390.0050 CA8.20390.0250
1,2-Diethoxyethane for synthesis, Sigma-Aldrich®
1,2-Diethoxyethane
Formula: C₆H₁₄O₂
MDL Number: MFCD00009253
CAS Number: 629-14-1
UN: 1153
ADR: 3,II

Order Now

Specification Test Results

Solubility 34 g/l (20 °C)
Melting Point -74 °C
Molar Mass 118.17 g/mol
Boiling Point 120 - 122 °C (1013 hPa)
Vapor Pressure 12.5 hPa (20 °C)
Flash Point 35 °C
Refractive Index 1.3922 (20 °C, 589 nm)
Density 0.84 g/cm3 (20 °C)
Ignition Point 208 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR