2,4,6-Triallyloxy-1,3,5-triazine stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: 2,4,6-Tris(allyloxy)-1,3,5-triazine, 2,4,6-Triallyloxy-s-triazine, 2,4,6-Tris(allyloxy)-s-triazine

M-Clarity™ quality level = MQ200

8.20359.0006 8.20359.0250 8.20359.0006 8.20359.0250
EM8.20359.0006CS 48322.72 CAD
EM8.20359.0006 EM8.20359.0250 CA8.20359.0006 CA8.20359.0250
2,4,6-Triallyloxy-1,3,5-triazine stabilized for synthesis, Sigma-Aldrich®
2,4,6-Triallyloxy-1,3,5-triazine
Formula: C₁₂H₁₅N₃O₃
MW: 249.27 g/mol
MDL Number: MFCD00006049
CAS Number: 101-37-1
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Solubility 0.3 g/l (20 °C)
Melting Point 26 - 27 °C
Molar Mass 249.27 g/mol
Boiling Point 149 - 150 °C (3 hPa) decomposes
Vapor Pressure 1.3 hPa (100 °C)
Flash Point 165 - 170 °C
Density 1.000 g/cm3 (20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR