2-tert-Butyl-4-methoxyphenol for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
3-BHA, 4-Hydroxy-3-tert-butylanisole, 3-tert-Butyl-4-hydroxyanisole
M-Clarity™ quality level = MQ200
8.20246.0100
8.20246.0500
8.20246.5000
8.20246.0100
8.20246.0500
8.20246.5000
EM8.20246.0100CS
794.74
CAD
EM8.20246.0100
EM8.20246.0500
EM8.20246.5000
CA8.20246.0100
CA8.20246.0500
CA8.20246.5000
2-tert-Butyl-4-methoxyphenol for synthesis, Sigma-Aldrich®
2-tert-Butyl-4-methoxyphenol
Formula:
C₁₁H₁₆O₂ |
MDL Number:
MFCD00040484 CAS Number: 121-00-6 |
Specification Test Results
Solubility | (20 °C) insoluble |
Melting Point | 48 - 63 °C |
Molar Mass | 180.25 g/mol |
Boiling Point | 263 °C (1013 hPa) |
Vapor Pressure | 1.33 hPa (105 °C) |
Flash Point | 130 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...