Sodium butyrate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
Synonyms:
Butyric acid sodium salt, Sodium Butyrate
M-Clarity™ quality level = MQ200
8.20236.0250
8.20236.1000
8.20236.0250
8.20236.1000
EM8.20236.0250CS
2997.26
CAD
EM8.20236.0250
EM8.20236.1000
CA8.20236.0250
CA8.20236.1000
Sodium butyrate for synthesis, Sigma-Aldrich®
Sodium butyrate
Formula:
C₄H₇NaO₂ MW: 110.09 g/mol |
MDL Number:
MFCD00002816 CAS Number: 156-54-7 |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | 244 °C |
Molar Mass | 110.09 g/mol |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...