di-Sodium succinate hexahydrate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate

M-Clarity™ quality level = MQ200

8.20151.0100 8.20151.0100 8.20151.0500
EM8.20151.0100CS 10246.3 CAD
EM8.20151.0100 CA8.20151.0100 CA8.20151.0500
di-Sodium succinate hexahydrate for synthesis, Sigma-Aldrich®
di-Sodium succinate hexahydrate
Formula: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Melting Pt: 300
MDL Number: MFCD00149117
CAS Number: 6106-21-4
Merck Index: 13,08754

Order Now

Specification Test Results

Solubility 349 g/l (25 °C)
Melting Point 120 °C Elimination of water of crystallisation
Molar Mass 270.15 g/mol
pH 6.0 - 8.5 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR