Aminoiminomethanesulfinic acid for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.20082.0250 8.20082.1000 8.20082.0250 8.20082.1000
EM8.20082.0250CS 2019.24 CAD
EM8.20082.0250 EM8.20082.1000 CA8.20082.0250 CA8.20082.1000
Aminoiminomethanesulfinic acid for synthesis, Sigma-Aldrich®
Aminoiminomethanesulfinic acid
Formula: CH₄N₂O₂S
MDL Number: MFCD00002397
CAS Number: 1758-73-2
UN: 3341
ADR: 4.2,II

Order Now

Specification Test Results

Solubility 27 g/l (20 °C)
Melting Point 123 °C (decomposition)
Molar Mass 108.12 g/mol
Bulk Density 850 kg/m3
Density 1.68 g/cm3 (20 °C)
pH 4 (10 g/l, H2O, 20 °C)
Hazard Class 4.2
Hazard Class Description Spontaneously Combustible Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR