Aluminium 99-100, granulated zS, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Aluminum

MQ200

8.20051.0100 8.20051.1000 8.20051.0100 8.20051.1000
EM8.20051.0100CS 3166.32 CAD
EM8.20051.0100 EM8.20051.1000 CA8.20051.0100 CA8.20051.1000
Aluminium 99-100, granulated zS, Sigma-Aldrich®
Aluminium
Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
MDL Number: MFCD00134029
CAS Number: 7429-90-5
Merck Index: 13,00321

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Melting Point 660 °C
Molar Mass 26.98 g/mol
Boiling Point 2467 °C
Density 2.70 g/cm3 (20 °C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR