Polyethylene glycol 3,000 for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
PEG 3,000
M-Clarity™ quality level = MQ200
8.19015.1000
8.19015.5000
8.19015.1000
8.19015.5000
EM8.19015.1000CS
763.14
CAD
EM8.19015.1000
EM8.19015.5000
CA8.19015.1000
CA8.19015.5000
Polyethylene glycol 3,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 3,000
Formula:
H(OCH₂CH₂)nOH MW: 3000 Density: 1.21 |
CAS Number:
25322-68-3 |
Specification Test Results
Solubility | 550 g/l (20 °C) |
Melting Point | 55 - 58 °C |
Molar Mass | 3000 g/mol |
Bulk Density | 400 - 500 kg/m3 |
Vapor Pressure |
|
Flash Point | 270 °C |
Density | 1.21 g/cm3 (20 °C) |
pH | 4 - 7 (100 g/l, H2O, 20 °C) |
Ignition Point | 420 °C |
Viscosity kinematic | 69 - 92 mm2/s (20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...