Polyethylene glycol 20,000 for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: PEG 20000, Ethylene glycol polymer 20000, PEG 20,000

M-Clarity™ quality level = MQ200

8.18897.1000 8.18897.5000 8.18897.1000 8.18897.5000
EM8.18897.1000CS 709.42 CAD
EM8.18897.1000 EM8.18897.5000 CA8.18897.1000 CA8.18897.5000
Polyethylene glycol 20,000 for synthesis, Sigma-Aldrich®
Polyethylene glycol 20,000
Formula: H(OCH₂CH₂)nOH
MW: 20000 g/mol
Flash Pt: 260
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Specification Test Results

Solubility 500 g/l (20 °C)
Melting Point 58 - 63 °C
Molar Mass 20000 g/mol
Bulk Density 400 - 500 kg/m3
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 260 °C
Density 1.2 g/cm3 (20 °C)
pH 4 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 320 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR