1-Amino-2-methoxyethane for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
2-Methoxyethylamine
M-Clarity™ quality level = MQ200
8.18872.0025
8.18872.0100
8.18872.0500
8.18872.2500
8.18872.0025
8.18872.0100
8.18872.0500
8.18872.2500
EM8.18872.0025CS
8129.1
CAD
EM8.18872.0025
EM8.18872.0100
EM8.18872.0500
EM8.18872.2500
CA8.18872.0025
CA8.18872.0100
CA8.18872.0500
CA8.18872.2500
1-Amino-2-methoxyethane for synthesis, Sigma-Aldrich®
1-Amino-2-methoxyethane
Formula:
C₃H₉NO MW: 75.11 g/mol |
MDL Number:
MFCD00008180 CAS Number: 109-85-3 UN: 2733 ADR: 3 (8),II |
Specification Test Results
Solubility | (20 °C) soluble |
Melting Point | -82 °C |
Molar Mass | 75.11 g/mol |
Boiling Point | 95 °C (1013 hPa) |
Flash Point | 12 °C |
Refractive Index | 1.4594 (20 °C, 589 nm) |
Explosion Limit | 3 - 16.10 %(V) |
Density | 0.87 g/cm3 (20 °C) |
pH | 12 (100 g/l, H2O, 20 °C) |
Ignition Point | 285 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...