Ethylene glycol dimethacrylate (ethylene dimethacrylate) stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

8.18847.0250 8.18847.1000 8.18847.0250 8.18847.1000
EM8.18847.0250CS 1415.68 CAD
EM8.18847.0250 EM8.18847.1000 CA8.18847.0250 CA8.18847.1000
Ethylene glycol dimethacrylate (ethylene dimethacrylate) stabilized for synthesis, Sigma-Aldrich®
Ethylene glycol dimethacrylate (Ethylene dimethacrylate)
Formula: C₁₀H₁₄O₄
MDL Number: MFCD00008590
CAS Number: 97-90-5

Order Now

Specification Test Results

Solubility  - 5 g/l (20 °C)
Melting Point -20 °C
Molar Mass 198.22 g/mol
Boiling Point 240 °C (1013 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 101.5 °C
Density 1.05 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR