Acetic acid 99 Acidimetry zS, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

MQ200

8.18755.2500 8.18755.1000 8.18755.2500 8.18755.9025
EM8.18755.2500CS 897.44 CAD
EM8.18755.2500 CA8.18755.1000 CA8.18755.2500 CA8.18755.9025
Acetic acid 99 Acidimetry zS, Sigma-Aldrich®
Acetic acid
Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 (lit.)
Melting Pt: 17 (lit.)
Density: 1.049 g/ml (25°C) (lit.)
Flash Pt: 38.5
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
UN: 2789
ADR: 8.3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric) ≥ 99.0 %
Density (d 20/4) 1.048 - 1.052
Identity passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR