Chlorotrimethylsilane for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: TMCS, TMSC, Trimethylchlorosilane

M-Clarity™ quality level = MQ200

8.18737.0100 8.18737.0500 8.18737.0100 8.18737.0500
EM8.18737.0100CS 1391.98 CAD
EM8.18737.0100 EM8.18737.0500 CA8.18737.0100 CA8.18737.0500
Chlorotrimethylsilane for synthesis, Sigma-Aldrich®
Chlorotrimethylsilane
Formula: C₃H₉ClSi
MDL Number: MFCD00000502
CAS Number: 75-77-4
UN: 1298
ADR: 3 (8),II

Order Now

Specification Test Results

Solubility (20 °C) (rigorous decomposition)
Melting Point -58 °C
Molar Mass 108.64 g/mol
Boiling Point 57 °C (1013 hPa)
Vapor Pressure 250 hPa (20 °C)
Flash Point -18 °C
Explosion Limit 1.5 - 46 %(V)
Density 0.86 g/cm3 (20 °C)
Ignition Point 420 °C
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR