2-Heptanone for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

8.18711.0100 8.18711.1000 8.18711.0100 8.18711.1000
EM8.18711.0100CS 677.96 CAD
EM8.18711.0100 EM8.18711.1000 CA8.18711.0100 CA8.18711.1000
2-Heptanone for synthesis, Sigma-Aldrich®
2-Heptanone
Formula: C₇H₁₄O
MDL Number: MFCD00009513
CAS Number: 110-43-0
UN: 1110
ADR: 3,III

Order Now

Specification Test Results

Solubility 4.3 g/l (20 °C)
Melting Point -35 °C
Molar Mass 114.18 g/mol
Boiling Point 150 - 152 °C (1013 hPa)
Vapor Pressure 4.5 hPa (20 °C)
Flash Point 43 °C
Refractive Index 1.4007 (20 °C, 589 nm)
Explosion Limit 1.11 - 7.9 %(V)
Density 0.82 g/cm3 (20 °C)
Ignition Point 532 °C
Saturation concentration 16.2 g/m3 (20 °C) Air
Evaporation number 17.4
Hazard Class 3
Hazard Class Description Flammable Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR