Formaldehyde 37% in aqueous solution stabilized for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


Stabilization: Stabilised with methanol 10%
M-Clarity™ quality level = MQ200

8.18708.5000 8.18708.1000 8.18708.5000
EM8.18708.5000CS 829.5 CAD
EM8.18708.5000 CA8.18708.1000 CA8.18708.5000
Formaldehyde 37% in aqueous solution stabilized for synthesis, Sigma-Aldrich®
Formaldehyde
Formula: CH₂O
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 2209
ADR: 8,III

Order Now

Specification Test Results

Density 1.017 g/cm3 (20 °C)
pH 0.5 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR