Chloramine-T (sodium salt) trihydrate for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, N-Chloro-4-toluenesulfonamide sodium salt, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
M-Clarity™ quality level = MQ200
8.18705.0005
8.18705.0250
8.18705.1000
8.18705.0005
8.18705.0250
8.18705.1000
EM8.18705.0005CS
66032.94
CAD
EM8.18705.0005
EM8.18705.0250
EM8.18705.1000
CA8.18705.0005
CA8.18705.0250
CA8.18705.1000
Chloramine-T (sodium salt) trihydrate for synthesis, Sigma-Aldrich®
Chloramine T (sodium salt) trihydrate
Formula:
CH₃C₆H₄SO₂NClNa·3H₂O |
MDL Number:
MFCD00149066 CAS Number: 7080-50-4 UN: 2928 ADR: 6.1 (8),II |
Specification Test Results
Solubility | 150 g/l (25 °C) |
Molar Mass | 281.69 g/mol |
Bulk Density | 540 - 680 kg/m3 |
Flash Point | 192 °C |
pH | 8 - 10 (50 g/l, H2O, 20 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...