Benzyl benzoate for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Benzoic acid benzyl ester

M-Clarity™ quality level = MQ200

8.18701.0100 8.18701.1000 8.18701.0100 8.18701.1000
EM8.18701.0100CS 900.6 CAD
EM8.18701.0100 EM8.18701.1000 CA8.18701.0100 CA8.18701.1000
Benzyl benzoate for synthesis, Sigma-Aldrich®
Benzyl benzoate
Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 323
Melting Pt: 21
Flash Pt: 158
MDL Number: MFCD00003075
CAS Number: 120-51-4
UN: 3082
ADR: 9 III
Merck Index: 13,01129

Order Now

Specification Test Results

Solubility 0.0153 g/l (20 °C)
Melting Point 21 °C
Molar Mass 212.24 g/mol
Boiling Point 324 °C (1013 hPa)
Vapor Pressure  - 0.1 hPa (20 °C)
Flash Point 158 °C
Density 1.12 g/cm3 (20 °C)
Ignition Point 480 °C
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR