Adipic acid zS, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
Hexanedioc acid, 1,6-Hexanedioic acid
MQ200
8.18650.0100
8.18650.1000
8.18650.0100
8.18650.1000
EM8.18650.0100CS
5259.82
CAD
EM8.18650.0100
EM8.18650.1000
CA8.18650.0100
CA8.18650.1000
Adipic acid zS, Sigma-Aldrich®
Adipic acid
Formula:
HOOC(CH₂)₄COOH MW: 146.14 g/mol Boiling Pt: 337 Melting Pt: 151 Flash Pt: 196 |
MDL Number:
MFCD00004420 CAS Number: 124-04-9 Merck Index: 13,00163 |
|
Specification Test Results
Assay (acidimetric) | ≥ 99.0 % |
Identity (IR) | passes test |
Melting range (lower value) | ≥ 150 °C |
Melting range (upper value) | ≤ 154 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...