Tributyl phosphate ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ200
8.18604.2500
8.18604.1000
8.18604.5000
8.18604.0100
8.18604.0500
8.18604.1000
8.18604.5000
CA8.18604.2500EA
759
CAD
CA8.18604.2500
EM8.18604.0100
EM8.18604.1000
EM8.18604.5000
CA8.18604.0100
CA8.18604.0500
CA8.18604.1000
CA8.18604.5000
Tributyl phosphate ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Tributyl phosphate
Formula:
(CH₃(CH₂)₃O)₃PO Boiling Pt: 180…183 °C/22 mmHg (lit.) Melting Pt: −79 °C (lit.) Density: 0.979 g/ml (25°C) (lit.) Flash Pt: 151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009436 CAS Number: 126-73-8 EINECS: 204-800-2 Merck Index: 13,09692 |
|
Specification Test Results
Assay (GC, area%) | ≥ 99.0 % (a/a) |
Density (d 20 °C/ 4 °C) | 0.976 - 0.978 |
Identity (IR) | passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...